Anti-MARCKS Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32590-50EA 882 EUR
ABCAAB32590-50
Anti-MARCKS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCKS (myristoylated alanine-rich protein kinase C substrate)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR