Anti-Gonadotropin Inducible Transcription Repressor 1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB32597-100EA 657 EUR
ABCAAB32597-100
Anti-Gonadotropin Inducible Transcription Repressor 1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Gonadotropin Inducible Transcription Repressor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR