Anti-GJB4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32598-100EA 672 EUR
ABCAAB32598-100
Anti-GJB4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GJB4 (gap junction protein, beta 4, 30.3kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR