Anti-Macrophage Inflammatory Protein 1 alpha / CCL3 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB32609-500EA 877 EUR
ABCAAB32609-500
Anti-Macrophage Inflammatory Protein 1 alpha / CCL3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Macrophage Inflammatory Protein 1 alpha / CCL3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human