Anti-F2R Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB32611-500EA 896 EUR
ABCAAB32611-500
Anti-F2R Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: F2R (coagulation factor II (thrombin) receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human