Anti-GRP78 BiP Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32618-500EA 697 EUR
ABCAAB32618-500
Anti-GRP78 BiP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRP78 BiP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR