Anti-RASGAP Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32625-100EA 634 EUR
ABCAAB32625-100
Anti-RASGAP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RASGAP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR