Anti-RNF20 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32629-100EA 713 EUR
ABCAAB32629-100
Anti-RNF20 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RNF20 (ring finger protein 20, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR