Anti-Mitochondrial dicarboxylate carrier Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32632-100EA 634 EUR
ABCAAB32632-100
Anti-Mitochondrial dicarboxylate carrier Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Mitochondrial dicarboxylate carrier
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR