Anti-CACNA1A Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB32642-100EA 688 EUR
ABCAAB32642-100
Anti-CACNA1A Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CACNA1A (calcium channel, voltage-dependent, P/Q type, alpha 1A subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR