Anti-RING1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32644-100EA 749 EUR
ABCAAB32644-100
Anti-RING1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RING1 (ring finger protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR