Anti-PPP2CB Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32673-100EA 666 EUR
ABCAAB32673-100
Anti-PPP2CB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPP2CB (protein phosphatase 2, catalytic subunit, beta isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR