Anti-HCN4 Rat monoclonal antibody unconjugated [clone: SHG 1E5]

Lieferant: Abcam
ABCAAB32675-100EA 703 EUR
ABCAAB32675-100
Anti-HCN4 Rat monoclonal antibody unconjugated [clone: SHG 1E5]
Antikörper
Type: Primary
Antigen: HCN4 (hyperpolarization activated cyclic nucleotide-gated potassium channel 4)
Clonality: monoclonal
Clone: SHG 1E5
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR