Anti-AKAP9 Mouse Monoclonal Antibody [clone: 17G10]

Lieferant: Abcam
ABCAAB32679-100EA 688 EUR
ABCAAB32679-100
Anti-AKAP9 Mouse Monoclonal Antibody [clone: 17G10]
Antikörper
Type: Primary
Antigen: AKAP9 (A kinase (PRKA) anchor protein 9)
Clonality: monoclonal
Clone: 17G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR