Anti-SULF1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32763-200EA 688 EUR
ABCAAB32763-200
Anti-SULF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SULF1 (sulfatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR