Anti-ACTN4 Mouse Monoclonal Antibody [clone: 7H6]

Lieferant: Abcam
ABCAAB32816-100EA 703 EUR
ABCAAB32816-100
Anti-ACTN4 Mouse Monoclonal Antibody [clone: 7H6]
Antikörper
Type: Primary
Antigen: ACTN4 (actinin, alpha 4)
Clonality: monoclonal
Clone: 7H6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR