Anti-TPH1 Sheep Polyclonal Antibody

Lieferant: Abcam
ABCAAB32821-50EA 672 EUR
ABCAAB32821-50
Anti-TPH1 Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPH1 (tryptophan hydroxylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR