Anti-PITX2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32832-100EA 672 EUR
ABCAAB32832-100
Anti-PITX2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PITX2 (paired-like homeodomain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR