Anti-CCDC50 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB32884-100EA 634 EUR
ABCAAB32884-100
Anti-CCDC50 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCDC50 (coiled-coil domain containing 50)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR