Anti-HTR2C Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB32887-100EA 672 EUR
ABCAAB32887-100
Anti-HTR2C Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTR2C (5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR