Anti-Nogo Receptor Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB32890-100EA 672 EUR
ABCAAB32890-100
Anti-Nogo Receptor Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nogo Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR