Anti-RICTOR Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB32892-100EA 896 EUR
ABCAAB32892-100
Anti-RICTOR Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RICTOR (RPTOR independent companion of MTOR, complex 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR