Anti-POMC Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB32893-100EA 944 EUR
ABCAAB32893-100
Anti-POMC Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR