Anti-APOE Goat polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB32897-100EA 882 EUR
ABCAAB32897-100
Anti-APOE Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human