Anti-CD2BP2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB32899-100EA 706 EUR
ABCAAB32899-100
Anti-CD2BP2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD2BP2 (CD2 (cytoplasmic tail) binding protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR