Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Abcam
ABCAAB32914-500EA 666 EUR
ABCAAB32914-500
Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen (Ad/Ay)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR