Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB32916-500EA 690 EUR
ABCAAB32916-500
Anti-Hepatitis B Virus Surface Antigen (Ad/Ay) Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Hepatitis B Virus Surface Antigen (Ad/Ay)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR