Anti-Bone Sialoprotein Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB33022-100EA 696 EUR
ABCAAB33022-100
Anti-Bone Sialoprotein Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Bone Sialoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR