Anti-SMG5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB33033-100EA 672 EUR
ABCAAB33033-100
Anti-SMG5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMG5 (SMG5 nonsense mediated mRNA decay factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR