Anti-SPP1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB33046-100EA 713 EUR
ABCAAB33046-100
Anti-SPP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SPP1 (secreted phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human