Anti-PKP3 Mouse Monoclonal Antibody [clone: E612B11F8]

Lieferant: Abcam
ABCAAB33070-100EA 666 EUR
ABCAAB33070-100
Anti-PKP3 Mouse Monoclonal Antibody [clone: E612B11F8]
Antikörper
Type: Primary
Antigen: PKP3 (plakophilin 3)
Clonality: monoclonal
Clone: E612B11F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR