Anti-NAP1L1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB33076-100EA 719 EUR
ABCAAB33076-100
Anti-NAP1L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NAP1L1 (nucleosome assembly protein 1-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR