Anti-FLT3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB33092-100EA 666 EUR
ABCAAB33092-100
Anti-FLT3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FLT3 (fms-related tyrosine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR