Anti-IgG Goat Polyclonal Antibody (Gold, 1.4 nm (ultra-small))

Lieferant: Abcam
ABCAAB33108-250EA 494 EUR
ABCAAB33108-250
Anti-IgG Goat Polyclonal Antibody (Gold, 1.4 nm (ultra-small))
Antikörper
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: 1.4 nm Gold (ultra-small)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR