Anti-alpha 1 Sodium Potassium ATPase Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB33133-50EA 666 EUR
ABCAAB33133-50
Anti-alpha 1 Sodium Potassium ATPase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: alpha 1 Sodium Potassium ATPase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR