Anti-CDH5 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB33168-100EA 738 EUR
ABCAAB33168-100
Anti-CDH5 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CDH5 (cadherin 5, type 2 (vascular endothelium))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR