Anti-EIF2S3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB33207-100EA 688 EUR
ABCAAB33207-100
Anti-EIF2S3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EIF2S3 (eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR