Anti-CD84 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: CD84.1.21]

Lieferant: Abcam
ABCAAB33217-50EA 666 EUR
ABCAAB33217-50
Anti-CD84 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: CD84.1.21]
Antikörper
Type: Primary
Antigen: CD84 (CD84 molecule)
Clonality: monoclonal
Clone: CD84.1.21
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR