Anti-PROX1 Mouse Monoclonal Antibody [clone: 5G10]

Lieferant: Abcam
ABCAAB33219-100EA 899 EUR
ABCAAB33219-100
Anti-PROX1 Mouse Monoclonal Antibody [clone: 5G10]
Antikörper
Type: Primary
Antigen: PROX1 (prospero homeobox 1)
Clonality: monoclonal
Clone: 5G10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR