Anti-HLA Class I Rat Monoclonal Antibody [clone: YTH862.2]

Lieferant: Abcam
ABCAAB33252-100EA 672 EUR
ABCAAB33252-100
Anti-HLA Class I Rat Monoclonal Antibody [clone: YTH862.2]
Antikörper
Type: Primary
Antigen: HLA Class I
Clonality: monoclonal
Clone: YTH862.2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR