Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TB01]

Lieferant: Abcam
ABCAAB33273-100EA 666 EUR
ABCAAB33273-100
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TB01]
Antikörper
Type: Primary
Antigen: B3GAT1 (beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P))
Clonality: monoclonal
Clone: TB01
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR