Anti-CD79B Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: AT107-2]

Lieferant: Abcam
ABCAAB33296-50EA 666 EUR
ABCAAB33296-50
Anti-CD79B Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: AT107-2]
Antikörper
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: monoclonal
Clone: AT107-2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR