Anti-MCAM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OJ79c]
Lieferant: Abcam
ABCAAB33300-50EA 876 EUR
ABCAAB33300-50
Anti-MCAM Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: OJ79c]
Antikörper
Type: Primary
Antigen: MCAM (melanoma cell adhesion molecule)
Clonality: monoclonal
Clone: OJ79c
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human