Anti-HIST3H3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB33309-100EA 929 EUR
ABCAAB33309-100
Anti-HIST3H3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR