Anti-CD151 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11G5a]

Lieferant: Abcam
ABCAAB33316-50EA 666 EUR
ABCAAB33316-50
Anti-CD151 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 11G5a]
Antikörper
Type: Primary
Antigen: CD151 (CD151 molecule (Raph blood group))
Clonality: monoclonal
Clone: 11G5a
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR