Anti-HLA B7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BB7.1]

Lieferant: Abcam
ABCAAB33331-50EA 666 EUR
ABCAAB33331-50
Anti-HLA B7 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BB7.1]
Antikörper
Type: Primary
Antigen: HLA B7
Clonality: monoclonal
Clone: BB7.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR