Anti-CD226 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: DX11]

Lieferant: Abcam
ABCAAB33335-50EA 337 EUR
ABCAAB33335-50
Anti-CD226 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: DX11]
Antikörper
Type: Primary
Antigen: CD226 (CD226 molecule)
Clonality: monoclonal
Clone: DX11
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR