Anti-CD226 Mouse Monoclonal Antibody (Biotin) [clone: DX11]

Lieferant: Abcam
ABCAAB33393-50EA 666 EUR
ABCAAB33393-50
Anti-CD226 Mouse Monoclonal Antibody (Biotin) [clone: DX11]
Antikörper
Type: Primary
Antigen: CD226 (CD226 molecule)
Clonality: monoclonal
Clone: DX11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR