Anti-CD226 Mouse Monoclonal Antibody [clone: DX11]
Lieferant: Abcam
ABCAAB33397-250EA 902 EUR
ABCAAB33397-250
Anti-CD226 Mouse Monoclonal Antibody [clone: DX11]
Antikörper
Type: Primary
Antigen: CD226 (CD226 molecule)
Clonality: monoclonal
Clone: DX11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human