Anti-CD3 Rat Monoclonal Antibody [clone: KT3]

Lieferant: Abcam
ABCAAB33429-125EA 697 EUR
ABCAAB33429-125
Anti-CD3 Rat Monoclonal Antibody [clone: KT3]
Antikörper
Type: Primary
Antigen: CD3
Clonality: monoclonal
Clone: KT3
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR